Description
门冬氨酸钾镁注射液含有的糖类一般为木糖醇或葡萄糖,可与与门冬氨酸分离。而安培检测器可进一步提高方法的选择性。经过分析柱选择和梯度条件优化,可使三种测定物实现较好分离,标准溶液谱图见图1。方法线性良好,灵敏度高,重现性好。| Market: | Pharma |
| Keywords: | 离子色谱;脉冲安培检测;CarboPac PA20;门冬氨酸;木糖醇;葡萄糖 |
| Matrix: | 注射液 |
For Research Use Only. Not for use in diagnostic procedures.