Find methods for your needs
HPLC-UV method for the rapid determination of alkaloids Be the first to rate this application
hplcuv method for rapid determination alkaloids
Description
A Thermo Scientific HPLC system is applied with the Thermo Scientific Syncronis aQ HPLC column for the analysis of alkaloids in less than three minutes with UV detection at 254 nm.
Market: Pharma
Keywords: Alkaloid, Anabasine, Cotinine, HPLC, Nicotine, Syncronis aQ, UV detection
Matrix: N/A
Author: Prakash Chander
Affiliation: Thermo Fisher Scientific
Uploaded on 10/28/2014.
For Research Use Only. Not for use in diagnostic procedures.