Find methods for your needs
Rapid separation of Raloxifene and its glucuronide metabolites from human plasma by LC-MS/MS Using SPE Micro Elution and a Thermo scientific Hypersil GOLD PFP column.
Description
The methods here demonstrate the advantages of using the Thermo Scientific Dionex UltiMate 3000 system; alongside the Thermo Scientific TSQ Vantage with a Thermo Scientific Hypersil GOLD PFP 3 μm column for the separation and determination of Raloxifene and its glucuronide metabolites from human plasma with MS detection. For Research Use Only. Not for use in diagnostic procedures.| Market: | Pharma |
| Keywords: | HPLC, Human Plasma, Hypersil GOLD PFP, MS detection, raloxifene, SOLAμ, SPE |
| Matrix: | Human plasma |
For Research Use Only. Not for use in diagnostic procedures.