Find methods for your needs
Related substances analysis of piracetam tablets according to the Chinese Pharmacopoeia using a Thermo Scientific Acclaim 120 C18 HPLC column
Description
In this work the related substances analysis of piracetam tablets is performed according to the Chinese Pharmacopoeia. The separation is performed using a Thermo Scientific Acclaim 120 C18 HPLC column with UV detection at 210 nm.| Market: | Pharma |
| Keywords: | Acclaim 120 C18, HPLC, Related substances, Tablet, UV detection, Chinese Pharmacopoeia, Piracetam |
| Matrix: | Tablet |
For Research Use Only. Not for use in diagnostic procedures.