Find methods for your needs
Rapid determination of Ethinyl Estradiol from human plasma using a Thermo Scientific Syncronis C18 HPLC column
Description
The Thermo Scientific Dionex UltiMate 3000 system is applied alongside the Thermo Scientific TSQ Vantage for the determination of ethinyl estradiol in human plasma. The sample was prepared using Thermo Scientific SOLA SCX SPE, followed by separation on a Thermo Scientific Syncronis C18 1.7 μm HPLC column. For Research Use Only. Not for use in diagnostic procedures.| Market: | Pharma |
| Keywords: | ethinyl estradiol, Human Plasma, LC-MS/MS, SOLA, SPE, Syncronis C18, UltiMate 3000 RSLC |
| Matrix: | Human plasma |
For Research Use Only. Not for use in diagnostic procedures.