Find methods for your needs
AB112: Determination of Organic Acids in Cranberry and Bilberry Extracts
Description
This application demonstrates the determination of quinic, malic, and citric acids in cranberry and bilberry extracts using a Thermo Scientific Dionex IonPac AS11-HC column with suppressed conductivity detection.| Market: | Food and Beverage |
| Keywords: | Dietary supplements, IonPac AS11-HC, standard reference materials, suppressed conductivity |
| Matrix: | Bilberry extracts, Cranberry |
For Research Use Only. Not for use in diagnostic procedures.