Find methods for your needs
Piracetam tablets assay according to the Chinese Pharmacopoeia using a Thermo Scientific Acclaim 120 C18 HPLC column
Description
In this work the assay of piracetam tablets according to the Chinese Pharmacopoeia is performed. The separation is performed using a Thermo Scientific Acclaim 120 C18 HPLC column with UV detection at 210 nm.Market: | Pharma |
Keywords: | Acclaim 120 C18, HPLC, Tablet, UV detection, Chinese Pharmacopoeia, Piracetam |
Matrix: | Tablet |
For Research Use Only. Not for use in diagnostic procedures.