Find methods for your needs
Fast analysis of Clortrimazole in athlete's foot ointment on a Thermo Scientific Acclaim 120 C8 HPLC column Be the first to rate this application
Description
The Thermo Scientific HPLC system is applied for the analysis of Clortrimazole in athlete's foot ointment. The separation is performed on a Thermo Scientific Acclaim 120 C8 HPLC column in under 5 minutes, with UV detection at 210 nm.
Market: Pharma
Keywords: Acclaim 120 C8, Athlete’s Foot Ointment, Clotrimazole, HPLC, UV detection
Matrix: Topical Ointment
Uploaded on 2/22/2010.
For Research Use Only. Not for use in diagnostic procedures.