Find methods for your needs
Separation of polymyxin sulfate B and its impurities in formulation using a simple isocratic HPLC-UV method
Description
The Thermo Scientific Dionex UltiMate 3000 system is applied for the impurity analysis of Polymyxin sulfate B. The separation was performed using a Thermo Scientific Syncronis C18 3 µm HPLC column with UV detection at 215 nm and a total run time of 60 minutes.| Market: | Pharma |
| Keywords: | Antibiotic, HPLC, Impurities, Polymyxin sulfate B, Syncronis C18, UV detection |
| Matrix: | N/A |
For Research Use Only. Not for use in diagnostic procedures.