Find methods for your needs
Fast UHPLC Method for the Simultaneous Determination of Free D-Aspartic Acid and D-Serine in Brain Tissue Extracts Be the first to rate this application
fast uhplc method for simultaneous determination free daspartic acid dserine brain tissue extracts
Description
The Thermo Scientific Dionex U3000 RSLC system is applied for a UHPLC method with automated precolumn derivatization using OPA-NAC and fluorescence detection for the nalysis of both D-Asp and D-Ser in brain tissue. Separation of D-and L-aspartic acid, D- and L- serine and other amino acids was done with a Thermo Scientific Hypersil Gold 1.9 μm column. With a total run time of the 21 minutes the UHPLC method faster analysis and enhanced resolution over typical HPLC methods.
Market: Other
Keywords: D-Aspartic Acid; D-Serine; Fluorescence Detection
Matrix: Brain tissue PCA extract
Uploaded on 10/17/2014.
For Research Use Only. Not for use in diagnostic procedures.