Description
本方法对比了单一色谱柱对肌醇的分析,发现因为饮料里含有许多在蒸发光散射检测器监测下有干扰的物质,通过常规的单一氨基柱对肌醇进行分析难以与杂质进行分离,所以选择一维氨基柱进行初步分离后,再在HILIC-1二维色谱柱上进行进一步的分离。该方法样品仅需简单过滤后,即可直接检测。| Market: | Food and Beverage |
| Keywords: | 在线二维;双三元;肌醇;功能饮料 |
| Matrix: | 饮料 |
For Research Use Only. Not for use in diagnostic procedures.