Description
本文采用带有疏水性固定相和弱阴离子交换官能团的混合分离模式色谱柱,成功实现了碘离子、硫氰酸根与样品中干扰峰的基线分离。为孕妇和婴幼儿奶粉中碘离子和硫氰酸根的检测提供了可行方法。| Market: | Food and Beverage |
| Keywords: | 碘离子;硫氰酸根;混合模式色谱柱;奶粉质量检测 |
| Matrix: | 奶粉 |
For Research Use Only. Not for use in diagnostic procedures.