Description
以甲苯为溶剂,提取母乳和奶粉中的脂肪酸,经乙酰氯-甲醇进行甲酯化后,用气相色谱-质谱联用仪测定脂肪酸甲酯的含量,全扫描方式采集数据,单个离子进行定量。结果表明该方法线性良好,方法简便快速。| Market: | Food and Beverage |
| Keywords: | 母乳;奶粉;脂肪酸;GC-MS(气相色谱-质谱法) |
| Matrix: | 乳类 |
For Research Use Only. Not for use in diagnostic procedures.