Description
本方法采用亲水液相色谱,使用U3000-CAD联用测定保健食品中左旋肉碱的含量,能使左旋肉碱获得较好保留,并且通过优化色谱条件使样品中杂质与左旋肉碱峰获得较好的分离。若对于实际样品测试时,应使用梯度方法以除去主峰后杂质对下一针样品的影响。而对于亲水色谱,梯度组成是水相比例由低到高能加强清洗能力。| Market: | Pharma |
| Keywords: | 亲水液相色谱;电喷雾检测器;左旋肉碱 |
| Matrix: | 左旋肉碱片 |
For Research Use Only. Not for use in diagnostic procedures.