Description
建立了一种简单易行的离子色谱方法,通过使用淋洗液在线发生装置,等度泵实现梯度分离,同时测定了电镀液中的甲酸、酒石酸、草酸、柠檬酸、SCN-、S2O32-等常见添加剂成分,这六种离子在23 min内可完全出峰,而共存的常见离子对此六种物质的分离都不存在干扰。与以往方法相比,本方法大大提高了待测组分的灵敏度,测定结果更为准确。快速的测定和准确的结果,使得电镀槽液的组分变化监测更为精确和实时化。| Market: | Chemical |
| Keywords: | 离子色谱;电导检测;IonPac AS19;电镀液;阴离子;有机酸 |
| Matrix: | 电镀液 |
For Research Use Only. Not for use in diagnostic procedures.