Description
本文采用KOH 梯度洗脱的离子色谱法,有效地分析测定了大气颗粒物中的水溶性无机阴离子氟离子,氯离子,硝酸根,硫酸根与小分子有机酸根甲酸根,乙酸根,草酸根,丙二酸根,丁二酸根及甲磺酸根。方法操作简单,检出限低,线性范围宽,并具有较好的重现性。对于实际样品中阴离子分析有较好的重现性和回收率,说明本方法有良好的适用性。| Market: | Environmental |
| Keywords: | 阴离子;大气颗粒物;电导检测;离子色谱 |
| Matrix: | 大气颗粒物 |
For Research Use Only. Not for use in diagnostic procedures.