Find methods for your needs
AN1062: Determination of Morpholine in Linezolid by Ion Chromatography
Description
Morpholine has been used in the production of many types of therapeutic agents, such as linezolid, a synthetic antobiotic of the oxazolidinone class. This application describes an IC method for the determination of low ug/L concentrations of morpholine in linezolid using a Reagent-Free IC (RFIC) system with suppressed conductivity detection.| Market: | Pharma |
| Keywords: | Antibiotics, Dionex IonPac CS19 Column, Dionex IonPac TCC-ULP1 Column, Suppressed Conductivity |
| Matrix: | 10% methanol |
| Author: | Lillian Chen, Brian De Borba, and Jeffrey Rohrer |
| Affiliation: | Thermo Fisher Scientific, Sunnyvale, CA USA |
For Research Use Only. Not for use in diagnostic procedures.