Find methods for your needs
AN1058: Determination of Polyacrylic Acid in Nuclear Power Plant Pressurized Water Reactor Secondary Feed Water
Description
Polyacrylic acid (PAA) has been used as a dispersant in the feed water to reduce the accumulation of metal oxide deposits on the secondary-side surfaces of the steam generators at nuclear power plants. This application describes a size-exclusion chromatography with UV detection method to determine low concentrations of PAA in secondary feed water containing ethanolamine (ETA) and hydrazine.| Market: | Water Analysis |
| Keywords: | Acclaim SEC-1000 Column, Corrosion Inhibition, Size-Exclusion Chromatography |
| Matrix: | Water |
| Author: | Lillian Chen, Brian Deborba, and Jeff Rohrer |
| Affiliation: | Thermo Fisher Scientific |
For Research Use Only. Not for use in diagnostic procedures.