Find methods for your needs
Separation of artemether and its impurities using reversed-phase HPLC-UV
Description
The Thermo Scientific Dionex UltiMate 3000 system is applied with the Thermo Scientific Accucore XL C18 HPLC column for the separation of artemether and its impurities, artemisine and dihydroartemisinin, using UV detection at 216 nm.| Market: | Pharma |
| Keywords: | Accucore XL C18, Antimalarial, Dihydroartemisinin, HPLC, UV detection, Artemisinin, Artemether |
| Matrix: | N/A |
| Author: | Anirudha Rakhe |
| Affiliation: | Thermo Fisher Scientific |
For Research Use Only. Not for use in diagnostic procedures.