Find methods for your needs
Aristolochic acids Be the first to rate this application
aristolochic acids
Description
Aristolochic acids in Aristolochia fangchi extract are separated within 15 min on a Thermo Scientific™ Accucore™ C18 column and detected by the Thermo Scientific™ Dionex™ Corona™ Veo™ RS charged aerosol detector.
Market: Food and Beverage
Keywords: Botanicals, Natural Products, Targeted Analysis, TCM Traditional Chinese Medicine, Universal Detection
Matrix: Methanolic extract
Uploaded on 10/10/2014.
For Research Use Only. Not for use in diagnostic procedures.