Description
Aristolochic acids in Aristolochia fangchi extract are separated within 15 min on a Thermo Scientific™ Accucore™ C18 column and detected by the Thermo Scientific™ Dionex™ Corona™ Veo™ RS charged aerosol detector.| Market: | Food and Beverage |
| Keywords: | Botanicals, Natural Products, Targeted Analysis, TCM Traditional Chinese Medicine, Universal Detection |
| Matrix: | Methanolic extract |
For Research Use Only. Not for use in diagnostic procedures.