Find methods for your needs
Rapid separation of triazine pesticides using a core enhanced technology C18 HPLC column
Description
A Thermo Scientific HPLC system was applied for the determination of triazine herbicides and pesticides. The separation was performed on a Thermo Scientific Accucore C18 column in less than 3 minutes, with UV detection at 280 nm.| Market: | Environmental |
| Keywords: | Accucore C18, Ametryn, Atrazine, Herbicides, HPLC, Pesticides, Prometryn, Propazine, Simazine, Simetryn, Triazines, UV detection |
| Matrix: | N/A |
| Author: | Derek Hillbeck |
| Affiliation: | Thermo Fisher Scientific |
For Research Use Only. Not for use in diagnostic procedures.