Find methods for your needs
Fast analysis of benzoic acids using a reversed-phase HPLC-UV method
Description
A Thermo Scientific HPLC system is applied with the Thermo Scientific Accucore 2.6 µm C18 column for the analysis of a range of benzoic acids in less than one minute with UV detection at 254 nm.| Market: | Pharma |
| Keywords: | 2-Fluorobenzoic acid, 3-Nitrobenzoic acid, Accucore C18, Benzoic acids, HPLC, m-Toluic acid, Phthalic acid, UV detection, Uracil, 3-Fluorobenzoic acid |
| Matrix: | N/A |
| Author: | Stephen Aspey |
| Affiliation: | Thermo Fisher Scientific |
For Research Use Only. Not for use in diagnostic procedures.