Find methods for your needs
Fast analysis of catecholamines using a solid core HILIC HPLC column
Description
The use of a Thermo Scientific Accucore HILIC column allowed the separation of seven catecholamine related molecules in less than five minutes at standard HPLC backpressures. Accucore HILIC columns are therefore an excellent choice for the fast analysis of catecholamines, allowing high sample throughput and analysis on conventional HPLC systems.| Market: | Pharma |
| Keywords: | 5-HIAA, Accucore HILIC, Catechol, Catecholamines, DOPAC, Hormones, L-DOPA, Neurotransmitters, Serotonin, Dopamine, L-tyrosine |
| Matrix: | N/A |
| Author: | Derek Hillbeck |
| Affiliation: | Thermo Fisher Scientific |
For Research Use Only. Not for use in diagnostic procedures.