Find methods for your needs
An improved separation of four antibiotics on a Thermo Scientific Acclaim 120 C18 HPLC column Be the first to rate this application
an improved separation four antibiotics on a thermo scientific acclaim 120 c18 hplc column
Description
The separation of four antibiotics was performed on a Thermo Scientific Acclaim 120 C18 5 µm column in under 15 minutes with UV detection.
Market: Pharma
Keywords: Acclaim 120 C18, Antibiotics, Chloramphenicol, HPLC, Oxytetracycline, Sulfadimethoxine, Sulfamerazine, UV detection
Matrix: N/A
Uploaded on 7/21/2014.
For Research Use Only. Not for use in diagnostic procedures.