Find methods for your needs
An improved analysis of flavonoids in green and black tea using a Thermo Scientific Acclaim 120 C18 HPLC column
Description
The separation of six flavonoids found in green and black tea was performed on a Thermo Scientific Acclaim 120 C18 3 µm column with UV detection.| Market: | Food and Beverage |
| Keywords: | Acclaim 120 C18, Black tea, Caffeine, Epicatechin, Epicatechin gallate, Epigallocatechin, Epigallocatechin gallate, Flavonoid, Green tea, HPLC, UV detection |
| Matrix: | Tea |
For Research Use Only. Not for use in diagnostic procedures.