Find methods for your needs
Improved assay of vanilla extract on a Thermo Scientific Acclaim 120 C18 HPLC column
Description
The Thermo Scientific Acclaim 120 C18 column is used for the analysis of vanilla extract. With an analysis time of under 16 minutes, the Thermo Scientific Acclaim 120 C18 column provides separation approximately 4 times faster than that by AOAC Official Method 990.25.| Market: | Food and Beverage |
| Keywords: | Acclaim 120 C18, AOAC 990.25, HPLC, UV detection, Vanilla extract, Vanillic acid, Vanillin |
| Matrix: | Vanilla extract |
For Research Use Only. Not for use in diagnostic procedures.