Find methods for your needs
Rapid determination of melamine in liquid milk and milk powder by HPLC on the Thermo Scientific Acclaim Mixed-Mode WCX-1 column
Description
The Thermo Scientific UltiMate 3000 HPLC system is applied for the analysis of melamine in liquid milk and milk powder. The separation is performed on a Thermo Scientific Acclaim Mixed-Mode WCX-1 HPLC column, with UV detection at 240 nm. The Thermo Scientific Acclaim Mixed-Mode WCX-1 column features a mixed-mode silica-based packing material that incorporates both hydrophobic and weak cation-exchange properties, and demonstrates great potential for separating samples that contain a mixture of ionic and neutral compounds.Market: | Food and Beverage |
Keywords: | Acclaim Mixed-Mode WCX-1, HPLC, melamine, milk, UV detection |
Matrix: | Milk |
For Research Use Only. Not for use in diagnostic procedures.