Find methods for your needs
Improved analysis of a strongly hydrophilic hydrotrope using a Thermo Scientific Acclaim Surfactant column Be the first to rate this application
improved analysis a strongly hydrophilic hydrotrope using a thermo scientific acclaim surfactant column
Description
The Thermo Scientific Acclaim Surfactant column is used for the analysis of a strongly hydrophilic hydrotrope, with UV detection at 225 nm. The weak anion-exchange properties of the Thermo Scientific Acclaim Surfactant column improve hydrotrope retention and resolution compared to purely reversed-phase columns.
Market: Pharma
Keywords: Acclaim Surfactant, HPLC, Hydrotropes, Sodium xylene sulfonate, Surfactant, UV
Matrix: N/A
Uploaded on 7/21/2014.
For Research Use Only. Not for use in diagnostic procedures.