Find methods for your needs
Accurate determination of the active ingredient ranitidine in Zantac 75 acid-reducer medication on a Thermo Scientific Acclaim 120 C18 HPLC column
Description
The Thermo Scientific HPLC system is applied for the determination of the active ingredient, ranitidine, in Zantac 75 acid-reducer medication. The separation is performed on a Thermo Scientific Acclaim 120 C18 HPLC column, with UV detection at 320 nm.Market: | Pharma |
Keywords: | Acclaim 120 C18, HPLC, Ranitidine, Ranitidine S-oxide, UV detection, Zantac |
Matrix: | Tablet |
For Research Use Only. Not for use in diagnostic procedures.