Find methods for your needs
Rapid analysis of pigments in turmeric on a Thermo Scientific Acclaim PolarAdvantage II (PA2) column
Description
The Thermo Scientific Acclaim PolarAdvantage II (PA2) 3 µm column is used to separate the three major yellow pigments known as curcuminoids in turmeric in less than 2.5 minutes.| Market: | Food and Beverage |
| Keywords: | Acclaim PolarAdvantage II (PA2), Bis-demethoxycurcumin, Curcumin, Demethoxycurcumin, HPLC, Pigment, Turmeric |
| Matrix: | Turmeric |
For Research Use Only. Not for use in diagnostic procedures.