Find methods for your needs
The effect of ionic strength on selectivity of a Thermo Scientific Acclaim Mixed-Mode WAX-1 HPLC column
Description
The Thermo Scientific Acclaim Mixed-Mode WAX-1 column is used to demonstrate the ionic strength effect on selectivity. Changing the ionic strength affects retention of anions without affecting neutral compounds.| Market: | Other |
| Keywords: | 4-Hydroxybenzoic acid, Acclaim Mixed-Mode WAX-1, Butylbenzene, HPLC, Ionic strength, UV detection |
| Matrix: | N/A |
For Research Use Only. Not for use in diagnostic procedures.