Find methods for your needs
The effect of ionic strength on selectivity of a Thermo Scientific Acclaim Mixed-Mode WAX-1 HPLC column Be the first to rate this application
effect ionic strength on selectivity a thermo scientific acclaim mixedmode wax1 hplc column
Description
The Thermo Scientific Acclaim Mixed-Mode WAX-1 column is used to demonstrate the ionic strength effect on selectivity. Changing the ionic strength affects retention of anions without affecting neutral compounds.
Market: Other
Keywords: 4-Hydroxybenzoic acid, Acclaim Mixed-Mode WAX-1, Butylbenzene, HPLC, Ionic strength, UV detection
Matrix: N/A
Uploaded on 7/17/2014.
For Research Use Only. Not for use in diagnostic procedures.