Find methods for your needs
Enhanced separation of alkylphenol ethoxylates on a Thermo Scientific Acclaim Surfactant column and a Thermo Scientific Acclaim Mixed-Mode HILIC-1 column
Description
Both the Thermo Scientific Surfactant and Thermo Scientific Acclaim Mixed-Mode HILIC-1 columns offer unique capabilities for the analysis of surfactants. The Acclaim Surfactant 5 µm column shows a complex profile that provides informtion on the distribution of ethoxylates. The Acclaim Mixed-Mode HILIC-1 column condenses the overlapping envelopes into two well-resolved peaks thus offering better quantification and detection limits.Market: | Material Science |
Keywords: | Acclaim Mixed-Mode HILIC-1, Acclaim Surfactant, Alkylphenol ethoxylates, Igepal CA-630, Igepal CO-630, Surfactant |
Matrix: | N/A |
For Research Use Only. Not for use in diagnostic procedures.