Find methods for your needs
Improved analysis of organic acids using a Thermo Scientific Acclaim Mixed-Mode WAX-1 and a Thermo Scientific Organic Acid column
Description
The Thermo Scientific Acclaim Mixed-Mode WAX-1 and Organic Acid columns are compared for the analysis of organic acids, with UV detection at 210nm. The unique chemistry of the Acclaim Mixed-Mode WAX-1 column cleanly separates compound which are challenging to separate using reversed-phase columns.| Market: | Food and Beverage |
| Keywords: | Acclaim Mixed-Mode WAX-1, Acclaim OA, Acetic acid, Ascorbic acid, Comparison, HPLC, Lactic acid, Organic acids |
| Matrix: | N/A |
For Research Use Only. Not for use in diagnostic procedures.