Find methods for your needs
Improved assay for pramoxine in topical anesthetic ointment on a Thermo Scientific Acclaim 120 C18 column with SPE clean-up
Description
The Thermo Scientific Acclaim 120 C18 column is applied for an assay for pramoxine in topical anesthetic ointment. SPE clean-up is performed using Thermo Scientific Dionex OnGuard II RP cartridge.| Market: | Pharma |
| Keywords: | Acclaim 120 C18, HPLC, OnGuard RP, Pramoxine, Reverse-phase, Solid phase extraction, SPE, Topical anesthetic ointment |
| Matrix: | Ointment |
For Research Use Only. Not for use in diagnostic procedures.