Find methods for your needs
Rapid determination of antimicrobial agents in hand soap using a Thermo Scientific Acclaim RSLC PolarAdvantage HPLC column
Description
The Thermo Scientific Dionex UltiMate 3000 RSLC system is applied for the determination of antimicrobial agents in hand soap. The separation is performed on a Thermo Scientific Acclaim RSLC PolarAdvtange (PA) column, with UV detection at 275 nm.| Market: | Pharma |
| Keywords: | Acclaim PolarAdvantage, Antimicrobial agents, HPLC, Isothiazolinones, Paraben, ProClin, Soap, UltiMate 3000 RSLC, UV detection |
| Matrix: | N/A |
For Research Use Only. Not for use in diagnostic procedures.