Find methods for your needs
Improved determination of diisocyanates by OSHA Method 42 on a Thermo Scientific Acclaim Mixed-Mode HILIC-1 as a confirmation column Be the first to rate this application
improved determination diisocyanates by osha method 42 on a thermo scientific acclaim mixedmode hilic1 as a confirmation column
Description
The Thermo Scientific Dionex UltiMate 3000 HPLC system is applied for the determination of diisocyanates by OSHA method 42. The confirmation analysis is performed on a Thermo Scientific Acclaim Mixed-Mode HILIC-1 column.
Market: Material Science
Keywords: 1,6-Hexamethylenediisocyanate, 2,4-Toluenediisocyanate, 2,6-Toluenediisocyanate, Acclaim Mixed-Mode HILIC-1, Diisocyanates, HPLC, OSHA method 42, UV detection
Matrix: N/A
Uploaded on 7/15/2014.
For Research Use Only. Not for use in diagnostic procedures.