Find methods for your needs
Fast analysis of azodicarbonamide on a Thermo Scientific Acclaim Mixed-Mode WAX-1 column in normal-phase mode
Description
The Thermo Scientific HPLC system is applied for the determination of azodicarbonamide. The analysis is performed on a Thermo Scientific Acclaim Mixed-Mode WAX-1 column, with UV detection at 242nm.| Market: | Material Science |
| Keywords: | Acclaim Mixed-Mode WAX-1, Azodicarbonamide, Blowing agent, HPLC, Normal-phase, UV detection |
| Matrix: | Azodicarbonamide |
For Research Use Only. Not for use in diagnostic procedures.