Find methods for your needs
Rapid determination of Carnitine in a nutritional supplement using a Thermo Scientific Acclaim Trinity P1 HPLC column
Description
The Thermo Scientific Dionex Ultimate 3000 RSLC system is applied for the analysis of carnitine and its counter-ion tartrate in a nutritional supplement. The separation was performed on a Thermo Scientific Acclaim Trinity P1 column in under 8 minutes, with UV detection at 210nm.| Market: | Pharma |
| Keywords: | Acclaim Trinity P1, Carnitine, HPLC, Tartrate, UltiMate 3000 RSLC, UV detection |
| Matrix: | Tablet dissolved in water |
For Research Use Only. Not for use in diagnostic procedures.