Find methods for your needs
Improved analysis of carotenoids and chlorophylls in vegetables using a Thermo Scientific Acclaim C30 column
Description
The Thermo Scientific Dionex UltiMate 3000 RS system is applied for the analysis of carotenoids and chlorophylls. The separation was performed in less than 25 minutes on a Thermo Scientific Acclaim C30 column with UV detection.| Market: | Food and Beverage |
| Keywords: | Acclaim C30, alpha-carotene, beta-carotene, beta-Cryptoxanthin, carotenoid, chlorophyll, Chlorophyll-a, Chlorophyll-b, Lutein, vegetables, Zeaxanthin |
| Matrix: | Carrot extracts in acetone, Maize, Spinach |
For Research Use Only. Not for use in diagnostic procedures.