Find methods for your needs
Rapid determination of Acetylcarnitine and Lipoic Acid in a nutritional supplement using a Thermo Scientific Acclaim Trinity P1 HPLC column
Description
The Thermo Scientific Dionex UltiMate 3000 RS system is applied for the analysis of acetylcarnitine and lipoic acid in nutritional supplements. The separation was performed on a Thermo Scientific Acclaim Trinity P1 column in under 8 minutes, with UV detection at 210 nm.| Market: | Pharma |
| Keywords: | Acclaim Trinity P1, Acetylcarnitine, HPLC, Lipoic Acid, Metabolism, UltiMate 3000 RS, UV detection |
| Matrix: | Tablet dissolved in water |
For Research Use Only. Not for use in diagnostic procedures.