Find methods for your needs
Rapid determination of Cimetidine using a Thermo Scientific Acclaim Trinity P1 HPLC column
Description
The Thermo Scientific Dionex Ultimate 3000 RSLC system is applied for the determination of cimetidine. The analysis was performed using a Thermo Scientific Acclaim Trinity P1 column with UV detection at 220 nm.| Market: | Pharma |
| Keywords: | Acclaim Trinity P1, Cimetidine, Histamine H2-receptor antagonist, HPLC, UV detection |
| Matrix: | Tablet |
For Research Use Only. Not for use in diagnostic procedures.