Find methods for your needs
Laureth Sulfate and Coco-amidopropyl Betaine in Shampoos Using Acclaim Surfactant Plus
Description
The Thermo Scientific Acclaim Surfactant Plus column is used to class-separate surfactants in two shampoo formulations with similar ingredient lists. The Thermo Scientific Dionex Corona Veo charged aerosol detector is used for these substances since they lack useful UV absorption.| Market: | Other |
| Keywords: | Acclaim, Corona, surfactant |
| Matrix: | Shampoo |
For Research Use Only. Not for use in diagnostic procedures.