Find methods for your needs
Sensitive and Rapid Determination of Polycyclic Aromatic Hydrocarbons in Tap Water
Description
This method describes an on-line SPE HPLC method with UV absorbance and fluorescence detections for rapid and sensitive determination of 20 PAHs in tap water. The determination is performed on a Thermo Scientific UltiMate 3000 Dual RSLC system combined with a Thermo Scientific Hypersil Green PAH analytical column. Reduced MDLs for UV and fluorescence detection, enabled by on-line SPE using the Thermo Scientific Dionex Acclaim PA2 column, provide a convenient method for determining these compounds in drinking and environmental waters using HPLC.| Market: | Water Analysis |
| Keywords: | Acclaim PA2 Column, HPLC, Hypersil Green PAH Column, On-Line SPE, Water Analysis |
| Matrix: | Tap water |
For Research Use Only. Not for use in diagnostic procedures.