Find methods for your needs
An improved determination of parabens in hand soap using the Thermo Scientific Acclaim RSLC PolarAdvantage HPLC column
Description
The Thermo Scientific Acclaim RSLC PolarAdvantage column was applied for the analysis of parabens in liquid hand soap. The separation of methyl and propyl paraben was achieved in less than five minutes with UV detection.| Market: | Pharma |
| Keywords: | Acclaim RSLC PolarAdvantage, Hand soap, HPLC, Methyl paraben, Preservatives, Propyl paraben, UV detection |
| Matrix: | Liquid hand soap |
For Research Use Only. Not for use in diagnostic procedures.