Find methods for your needs
Demonstration of EPA Method 555 to separate chlorinated acids in drinking water using a Thermo Scientific Acclaim RSLC PolarAdvantage HPLC column Be the first to rate this application
Description
EPA Method 555 is a HPLC method for the determination of certain chlorinated acids in ground water and finished drinking water using reversed-phase columns and diode-array detection. In this example, a fast, high-resolution separation of chlorinated acids in drinking water is achieved using the Thermo Scientific Acclaim RSLC PolarAdvantage 2.2 µm column with an analysis cycle time of about 12 minutes.
Market: Environmental
Keywords: 2,4,5-Trichlorophenoxyacetic acid (2,4,5-T), 2-methyl-4-chlorophenoxyacetic acid (MCPA), 3,5-Dichlorobenzoic acid, 4-(2,4-dichlorophenoxy)butyric acid (2,4-DB), 4-Nitrophenol, Chlorinated acids, Dinoseb, EPA Method 555, HPLC, Methylchlorophenoxypropionic acid (MCPP), Pentachlorophenol, UV detection
Matrix: Drinking water
Uploaded on 6/19/2014.
For Research Use Only. Not for use in diagnostic procedures.