Description
The Thermo Scientific Acclaim Surfactant Plus column can provide a detailed profile of the variation in ethoxylation number of the alkylphenolethoxylate family of surfactants. In this work a simple isocratic method is used to provide a detailed profile of Triton X-100 which is a prime example of this type of surfactant.| Market: | Chemical |
| Keywords: | HPLC, Acclaim Surfactant Plus, Triton X-100, UV detection, Alkylphenolethoxylate surfactants |
| Matrix: | N/A |
For Research Use Only. Not for use in diagnostic procedures.