Find methods for your needs
Simple isocratic separation of acetaldehyde, lactic acid and acetic acid using a mixed-mode HPLC column
Description
In this work, the separation of acetaldehyde, lactic acid and acetic acid was achieved using the Thermo Scientific Acclaim Mixed-Mode WAX-1 column. This column allows control over two retention mechanisms, leading to the development of a simple isocratic separation method.| Market: | Chemical |
| Keywords: | Acclaim Mixed-Mode WAX-1, Acetaldehyde, Acetic acid, HPLC, Lactic acid, UV detection |
| Matrix: | N/A |
For Research Use Only. Not for use in diagnostic procedures.