Description
This application describes the determination of cations and ammonium in drinking and waste water by suppressed ion chromatography as per ASTM method D6919-09. Non-mandatory water quality standards have been established by the U.S. Environmental Protection Agency (EPA) for these contaminants, whcih influnence the aesthetic character of drinking water. An isocratic method was used with a Dionex IonPac CS16 column on a Thermo Scientific Dionex ICS-5000 IC system using eluent generation.| Market: | Environmental |
| Matrix: | Drinking water, Waste water |
For Research Use Only. Not for use in diagnostic procedures.